Skip to Content
Merck
All Photos(3)

Key Documents

165956

Sigma-Aldrich

Methyl benzenesulfonate

98%

Synonym(s):

Benzenesulfonic acid methyl ester, Benzenesulfonic acid, methyl ester, Methyl benzenesulphonate, Methyl besylate, methyl ester of benzenesulphonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$191.00
500 G
$580.00

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$191.00
500 G
$580.00

About This Item

Empirical Formula (Hill Notation):
C7H8O3S
CAS Number:
Molecular Weight:
172.20
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

functional group

sulfonic acid

storage temp.

2-8°C

SMILES string

COS(=O)(=O)c1ccccc1

InChI

1S/C7H8O3S/c1-10-11(8,9)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

CZXGXYBOQYQXQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl benzenesulfonate can be used asa methylation agent:
  • For the C-methylation of perbromothiophene tosynthesize tribromo-methyl thiophene in the presence of butyllithium.
  • In the nickel-catalyzedmethylation of unactivated alkyl halides and acid chlorides to prepare methylatedalkanes and ketones.
It can alsobe used as a reactant in the etherification of sophoridinol derivatives tosynthesize N-substituted sophoridinic methyl ether derivatives.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julian L Goggi et al.
Molecular imaging and biology, 22(4), 993-1002 (2020-02-02)
Cancer immunotherapy has shown huge potential in the fight against cancer, but only a small proportion of patients respond successfully to treatment. Non-invasive methods to stratify responders from non-responders are critically important as immune therapies are often associated with immune-related
Zhen Liu et al.
Journal of pharmaceutical and biomedical analysis, 168, 23-29 (2019-02-21)
Alkyl sulfonate esters have been widely concerned as genotoxic impurities (GTIs). A gas chromatography triple-quadrupole mass spectrometry (GC-MS/MS) method has been developed for trace determination of 9 commonly encountered sulfonate esters in drug substances. Three different solvents of acetonitrile (ACN)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service