Skip to Content
Merck
All Photos(1)

Key Documents

163813

Sigma-Aldrich

Ethyl fluoroacetate

98%

Synonym(s):

Ethyl 2-fluoroacetate, Fluoroacetic acid ethyl ester, Monofluoroacetic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$79.80
50 G
$123.00

$79.80


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$79.80
50 G
$123.00

About This Item

Linear Formula:
FCH2COOC2H5
CAS Number:
Molecular Weight:
106.10
Beilstein:
1743761
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$79.80


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.375 (lit.)

bp

119.3 °C/753 mmHg (lit.)

solubility

water: soluble

density

1.098 g/mL at 25 °C (lit.)

functional group

ester
fluoro

SMILES string

CCOC(=O)CF

InChI

1S/C4H7FO2/c1-2-7-4(6)3-5/h2-3H2,1H3

InChI key

VCYZVXRKYPKDQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl fluoroacetate is an ester organic solvent[1].

Application

Ethyl fluoroacetate was used in the synthesis of 4-deoxy-4-fluoro-muscarines[2]. It was used as starting reagent for the synthesis of ethyl(diethoxyphosphoryl)fluoroacetate[3]. It was used in development of an ammonolysis-based microencapsulation process[1].

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A Convenient Synthesis of Ethyl(Diethoxyphosphoryl) fluoroacetate from Ethyl Fluoroacetate.
Elkik E and Imbeaux M.
Synthesis, 1989(11), 861-862 (1989)
P Bravo et al.
Journal of medicinal chemistry, 35(17), 3102-3110 (1992-08-21)
Four isomers of [(4-fluoro-5-methyl-tetrahydrofuran-2-yl)methyl]trimethylammonium iodide (4-deoxy-4-fluoro-muscarines) were prepared in enantiomerically and diastereomerically pure form from (S)-(-)-methyl 4-methylphenyl sulfoxide, ethyl fluoroacetate, and allyl bromide. Their absolute configurations were assigned by 1H NMR analyses. The four optically pure compounds were tested in
Reactivity of ethyl acetate and its derivatives toward ammonolysis: ramifications for ammonolysis-based microencapsulation process.
Chung Y, et al.
Polymers For Advanced Technologies, 20(10), 785-794 (2009)
Youngwook Park et al.
The journal of physical chemistry letters, 9(15), 4282-4286 (2018-07-13)
Chemical reactions are extremely difficult to occur in ice at low temperature, where atoms and molecules are frozen in position with minimal thermal energy and entropy. Contrary to this general behavior, certain weak acids including fluoroacetic acids dissociate spontaneously and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service