Skip to Content
Merck
All Photos(1)

Key Documents

163228

Sigma-Aldrich

2-Methyl-1-tetralone

98%

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 G
$226.00

$226.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2.5 G
$226.00

About This Item

Empirical Formula (Hill Notation):
C11H12O
CAS Number:
Molecular Weight:
160.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$226.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.5535 (lit.)

bp

127-131 °C/12 mmHg (lit.)

density

1.057 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC1CCc2ccccc2C1=O

InChI

1S/C11H12O/c1-8-6-7-9-4-2-3-5-10(9)11(8)12/h2-5,8H,6-7H2,1H3

InChI key

GANIBVZSZGNMNB-UHFFFAOYSA-N

General description

2-Methyl-1-tetralone undergoes enantioselective hydrogenation catalyzed by 1,4-diamine-ruthenium(II) complexes[1].

Application

2-Methyl-1-tetralone was used in enantioselective separation of indan, tetralin and benzosuberan derivatives in the presence of chiral additives by capillary electrophoresis[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takeshi Ohkuma et al.
Organic letters, 6(16), 2681-2683 (2004-07-30)
A combined system of a RuCl(2)(binap)(1,4-diamine) complex and t-C(4)H(9)OK in i-C(3)H(7)OH catalyzes enantioselective hydrogenation of various 1-tetralone derivatives and some methylated 2-cyclohexenones. Hydrogenation of 2-methyl-1-tetralone under dynamic kinetic resolution gives the cis alcohol with high ee. [reaction: see text]
Chiral separations of indan, tetralin and benzosuberan derivatives by capillary electrophoresis.
Gahm K-H, et al.
Journal of Chromatography A, 793(1), 135-143 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service