Skip to Content
Merck
All Photos(1)

Key Documents

162647

Sigma-Aldrich

Z-Gln-OH

99%

Synonym(s):

N-Cbz-L-glutamine, Carbobenzyloxy-L-glutamine, Z-L-Glutamine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$121.00

$121.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$121.00

About This Item

Linear Formula:
NH2COCH2CH2CH(NHCOOCH2C6H5)COOH
CAS Number:
Molecular Weight:
280.28
Beilstein:
2061271
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$121.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

99%

form

powder

optical activity

[α]23/D −7.3°, c = 2 in ethanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

134-138 °C (lit.)

application(s)

peptide synthesis

SMILES string

NC(=O)CC[C@H](NC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C13H16N2O5/c14-11(16)7-6-10(12(17)18)15-13(19)20-8-9-4-2-1-3-5-9/h1-5,10H,6-8H2,(H2,14,16)(H,15,19)(H,17,18)/t10-/m0/s1

InChI key

JIMLDJNLXLMGLX-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service