Skip to Content
Merck
All Photos(1)

Key Documents

162442

Sigma-Aldrich

2-Chloro-4-fluorophenol

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$140.00

$140.00

PRICE WITHOUT NATIONAL TAXES

0 left in stock

Request a Bulk Order

Select a Size

Change View
25 G
$140.00

About This Item

Linear Formula:
ClC6H3(F)OH
CAS Number:
Molecular Weight:
146.55
Beilstein:
1861277
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$140.00

PRICE WITHOUT NATIONAL TAXES

0 left in stock

Request a Bulk Order

Assay

99%

form

solid

refractive index

n20/D 1.53 (lit.)

bp

88 °C/4 mmHg (lit.)

mp

23 °C (lit.)

density

1.344 g/mL at 25 °C (lit.)

functional group

chloro
fluoro

SMILES string

Oc1ccc(F)cc1Cl

InChI

1S/C6H4ClFO/c7-5-3-4(8)1-2-6(5)9/h1-3,9H

InChI key

IGYXYGDEYHNFFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloro-4-fluorophenol was used in enzymatic production of fluorocatechols[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inna P Solyanikova et al.
Applied and environmental microbiology, 69(9), 5636-5642 (2003-09-06)
The present study describes the (19)F nuclear magnetic resonance analysis of the conversion of 3-halocatechols to lactones by purified chlorocatechol 1,2-dioxygenase (ClcA2), chloromuconate cycloisomerase (ClcB2), and chloromuconolactone dehalogenase (ClcF) from Rhodococcus opacus 1cp grown on 2-chlorophenol. The 3-halocatechol substrates were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service