Skip to Content
Merck
All Photos(1)

Key Documents

16175

Sigma-Aldrich

2-(2-Bromoethyl)-2,5,5-trimethyl-1,3-dioxane

technical, ≥90.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17BrO2
CAS Number:
Molecular Weight:
237.13
Beilstein:
4305359
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90.0% (AT)

form

liquid

refractive index

n20/D 1.470 (lit.)

bp

72-73 °C/0.6 mmHg (lit.)

density

1.26 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)COC(C)(CCBr)OC1

InChI

1S/C9H17BrO2/c1-8(2)6-11-9(3,4-5-10)12-7-8/h4-7H2,1-3H3

InChI key

HSWLDQSQVWEZIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Masked 2-bromoethyl methyl ketone forming a stable Grignard reagent

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reductive Cyclization of (3-) and (4-Oxoalkyl)-9, 10-anthraquinones to the cyclopenta [a] anthraquinone and naphthacene-5, 12-dione systems.
Krohn K and Bernhard S.
J. Prakt. Chem./Chem.-Ztg., 340(1), 26-33 (1998)
The Total synthesis of (+)-artemisinin and (+)-9-desmethyltemesinin.
Avery MA, et al.
Tetrahedron Letters, 28(4), 4629-4632 (1987)
W Francke et al.
Bioorganic & medicinal chemistry, 4(3), 363-374 (1996-03-01)
Head-space volatiles obtained from male mountain pine beetles, Dendroctonus ponderosae, were analyzed by coupled GC-MS and chiral gas chromatography. 5-Ethyl-7-methyl-6,8-dioxabicyclo[3.2.1]octane (6) was found as a new naturally occurring isomer of brevicomin (1). In addition, several stereoisomers of 7-ethyl-5-methyl-6,8-dioxabicyclo[3.2.1]octan-2-ol (11) and
J.C. Stowell et al.
Organic Syntheses, 62, 140-140 (1984)
B.B. Snider et al.
The Journal of Organic Chemistry, 49, 153-153 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service