161535
N-(2-Hydroxyethyl)ethylenediaminetriacetic acid trisodium salt hydrate
99%
Synonym(s):
Trisodium HEDTA hydrate
Sign Into View Organizational & Contract Pricing
Select a Size
100 G
$52.30
1 KG
$210.00
3 KG
$441.00
All Photos(3)
Select a Size
Change View
100 G
$52.30
1 KG
$210.00
3 KG
$441.00
About This Item
Linear Formula:
HOCH2CH2N(CH2CO2Na)CH2CH2N(CH2CO2Na)2 · xH2O
CAS Number:
Molecular Weight:
344.20 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
Assay
99%
form
solid
mp
288 °C (dec.) (lit.)
functional group
amine
SMILES string
O.[Na+].[Na+].[Na+].OCCN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
InChI
1S/C10H18N2O7.3Na.H2O/c13-4-3-11(5-8(14)15)1-2-12(6-9(16)17)7-10(18)19;;;;/h13H,1-7H2,(H,14,15)(H,16,17)(H,18,19);;;;1H2/q;3*+1;/p-3
InChI key
FHGJYTNPLCNRLG-UHFFFAOYSA-K
Application
N-(2-Hydroxyethyl)ethylenediaminetriacetic acid trisodium salt hydrate can be used as a complexing agent in combination with thiourea and its derivatives to enhance the deposition rate of electroless copper plating solutions.[1]
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The acceleration of nonformaldehyde electroless copper plating
Li J and Kohl PA
Journal of the Electrochemical Society, 149(12), C631-C636 (2002)
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service