Skip to Content
Merck
All Photos(1)

Key Documents

16005

Sigma-Aldrich

Paraformaldehyde

meets analytical specification of DAC, 95.0-100.5%

Synonym(s):

Polyoxymethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2O)nH
CAS Number:
Molecular Weight:
30.03 (as monomer)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
technique(s):
NMR: suitable
vapor pressure:
<1.45 mmHg ( 25 °C)
Pricing and availability is not currently available.

vapor density

1.03 (vs air)

vapor pressure

<1.45 mmHg ( 25 °C)

Assay

95.0-100.5%

form

pellets

autoignition temp.

572 °F

quality

meets analytical specification of DAC

expl. lim.

73 %

technique(s)

NMR: suitable

impurities

residual solvents, in accordance
<0.1% sulfated ash
10 ppm heavy metals (as Pb)

mp

120-170 °C (lit.)

solubility

water: insoluble

density

0.88 g/mL at 25 °C (lit.)

suitability

in accordance for appearance of solution
suitable for acidity or alkalinity (complying)

storage temp.

2-8°C

SMILES string

[O-]#[C+H2]

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Buffered picric acid-paraformaldehyde fixative is suitable for use in electron microscopy and light-microscopic immunocytochemistry.[1]

Application

Paraformaldehyde was used in the preparation of azomethine ylide, required for the synthesis of amine-modifying single-walled carbon nanotubes.[2] It was also used in the preparation of α-methylenechromanes, α-methylenequinoline and ortho-quinone methides.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Flam. Sol. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Justin Mulvey et al.
International journal of nanomedicine, 9, 4245-4255 (2014-09-18)
We aimed to create a more robust and more accessible standard for amine-modifying single-walled carbon nanotubes (SWCNTs). A 1,3-cycloaddition was developed using an azomethine ylide, generated by reacting paraformaldehyde and a side-chain-Boc (tert-Butyloxycarbonyl)-protected, lysine-derived alpha-amino acid, H-Lys(Boc)-OH, with purified SWCNT
P Somogyi et al.
Neuroscience, 7(7), 1779-1783 (1982-07-01)
The buffered picric acid paraformaldehyde fixative originally recommended for electronmicroscopy and which has since been used occasionally for light-microscopic immunocytochemistry, has been supplemented with glutaraldehyde and used as primary fixative for the perfusion of rat brains. In the basal ganglia
José C J M D S Menezes et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(42), 13644-13655 (2014-08-30)
In view of increasing demands for efficient photosensitizers for photodynamic therapy (PDT), we herein report the synthesis and photophysical characterizations of new chlorin e6 trimethyl ester and protoporphyrin IX dimethyl ester dyads as free bases and Zn(II) complexes. The synthesis of these
Lingling Chen et al.
Scientific reports, 7(1), 12218-12218 (2017-09-25)
Optical clearing methods can facilitate deep optical imaging in biological tissue by reducing light scattering and this has enabled accurate three-dimensional signal visualization and quantification of complex biological structures. Unfortunately, existing optical clearing approaches present a compromise between maximizing clearing
Ting Yao et al.
FEBS open bio, 6(7), 707-719 (2016-07-12)
Decorin (DCN) is a major member of the small leucine-rich proteoglycan (SLRP) family that is critically involved in tumorigenesis and the development of metastasis of cancers, including glioma. Overexpression of DCN was indicated to suppress glioma cell growth. However, the

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service