Skip to Content
Merck
All Photos(1)

Key Documents

157783

Sigma-Aldrich

Phosphorus tribromide

97%

Synonym(s):

Phosphorus(III) bromide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$136.00
500 G
$235.00

$136.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$136.00
500 G
$235.00

About This Item

Linear Formula:
PBr3
CAS Number:
Molecular Weight:
270.69
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$136.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor pressure

0.27 psi ( 54 °C)

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.697 (lit.)

bp

175 °C (lit.)

mp

−41.5 °C (lit.)

density

2.88 g/mL at 20 °C (lit.)

SMILES string

BrP(Br)Br

InChI

1S/Br3P/c1-4(2)3

InChI key

IPNPIHIZVLFAFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphorus tribromide (PBr3) is commonly used in Hell-Volhard-Zelinsky halogenation for the a-bromination of carboxylic acids to form the corresponding acyl bromide. It is also useful for the conversion of primary and secondary alcohols to alkyl bromides.[1][2]

Application

Phosphorus tribromide may be used as a reagent:
  • In a novel protocol for synthesizing 7-ethoxy-1-(p-ethylphenoxy)-3,7-dimethyl-2-octene, a synthetic juvenile hormone mimic of trans,trans,cis-10,11-epoxy-7-ethyl-3,11-dimethyltrideca-2,6-dienoate.[3]
  • To synthesize 1,2,3-diazaphosphinane, 1,2,3-thiazaphosphinine and 1,2-azaphosphole bearing a chromone ring.[4]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Chemistry, 1 (1988)
Organic Chemistry (2008)
Reaction of 2-cyano-3-(4-oxo-4H-chromen-3-yl) prop-2-enamide with some phosphorus reagents: synthesis of some novel diethyl phosphonates, 1, 2, 3-diazaphosphinanes, 1, 2, 3-thiazaphosphinine and 1, 2-azaphospholes bearing a chromone ring.
Ali TE & Hassan MM
Research on Chemical Intermediates, 44(1), 173-189 (2018)
Phosphorus tribromide promoted allylic rearrangement of a tertiary vinyl carbinol. Stereochemistry of the reaction product and application to the synthesis of JH-25, a potent juvenile hormone mimic.
Babler JH
The Journal of Organic Chemistry, 41(7), 1262-1264 (1976)
Andrey S Mereshchenko et al.
Nature chemistry, 7(7), 562-568 (2015-06-24)
'Roaming' is a new and unusual class of reaction mechanism that has recently been discovered in unimolecular dissociation reactions of isolated molecules in the gas phase. It is characterized by frustrated bond cleavage, after which the two incipient fragments 'roam'

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service