Skip to Content
Merck
All Photos(1)

Key Documents

157554

Sigma-Aldrich

1-Bromo-2,4-dimethoxybenzene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$45.10

$45.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$45.10

About This Item

Linear Formula:
BrC6H3(OCH3)2
CAS Number:
Molecular Weight:
217.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$45.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.572 (lit.)

bp

153-155 °C/18 mmHg (lit.)

mp

25-26 °C (lit.)

density

1.507 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

COc1ccc(Br)c(OC)c1

InChI

1S/C8H9BrO2/c1-10-6-3-4-7(9)8(5-6)11-2/h3-5H,1-2H3

InChI key

NIUZVSQOXJIHBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Bromo-2,4-dimethoxybenzene was used in the synthesis of 2,3-disubstituted benzo[b]furans.[1] It was also used in the synthesis of dendrimer Si[CH2CH2Si(Me)2-2,4-(MeO)2-C6H3]4.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dawei Yue et al.
The Journal of organic chemistry, 70(25), 10292-10296 (2005-12-06)
[reaction: see text] 2,3-Disubstituted benzo[b]furans are readily prepared under very mild reaction conditions by the palladium/copper-catalyzed cross-coupling of various o-iodoanisoles and terminal alkynes, followed by electrophilic cyclization with I2, PhSeCl, or p-O2NC6H4SCl. Aryl- and vinylic-substituted alkynes undergo electrophilic cyclization in
Selective lithiation and crystal structures of G1-carbosilane dendrimers with dimethoxybenzene functionalities.
Harder S, et al.
Journal of Organometallic Chemistry, 689(7), 1095-1101 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service