Skip to Content
Merck
All Photos(2)

Key Documents

157201

Sigma-Aldrich

Sodium aurothiomalate hydrate

Synonym(s):

(1,2-Dicarboxyethylthio)gold disodium salt hydrate, Gold sodium thiomalate hydrate, Mercaptobutanedioic acid gold(I) disodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$216.00
1 G
$591.00

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$216.00
1 G
$591.00

About This Item

Linear Formula:
NaO2CCH2CH(SAu)CO2Na · xH2O
Molecular Weight:
390.08 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

SMILES string

O.[Na+].[Na+].[O-]C(=O)CC(S[Au])C([O-])=O

InChI

1S/C4H6O4S.Au.2Na.H2O/c5-3(6)1-2(9)4(7)8;;;;/h2,9H,1H2,(H,5,6)(H,7,8);;;;1H2/q;3*+1;/p-3

InChI key

YLQOAPBVYJCTPW-UHFFFAOYSA-K

General description

Sodium aurothiomalate has antirheumatic properties and exhibits inhibitory effects against M. tuberculosis.[1]

Application

Sodium aurothiomalate hydrate was used in the high mobility group box chromosomal protein 1 (HMGB1) western blotting.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecilia K Zetterström et al.
Journal of leukocyte biology, 83(1), 31-38 (2007-10-05)
Gold compounds such as gold sodium thiomalate (GST) can reduce the symptoms of rheumatoid arthritis (RA), although their mechanism of action is not well defined. As the proinflammatory mediator high mobility group box chromosomal protein 1 (HMGB1) may play a
Kathryn E A Lougheed et al.
Tuberculosis (Edinburgh, Scotland), 89(5), 364-370 (2009-08-25)
Mycobacterium tuberculosis has an on-going impact on global public health and new therapeutics to treat tuberculosis are urgently required. The emergence of drug resistant tuberculosis poses a serious threat to the control of this pathogen, and the development of drugs
Christoph Tabeling et al.
Journal of vascular research, 52(4), 279-288 (2016-02-20)
Pulmonary vascular hyperresponsiveness is a main characteristic of pulmonary arterial hypertension (PAH). In PAH patients, elevated levels of the vasoconstrictors thromboxane A2 (TXA2), endothelin (ET)-1 and serotonin further contribute to pulmonary hypertension. Protein kinase C (PKC) isozyme alpha (PKCα) is
Meghan Riddell et al.
Nature communications, 9(1), 5357-5357 (2018-12-19)
Strict regulation of proliferation is vital for development, whereas unregulated cell proliferation is a fundamental characteristic of cancer. The polarity protein atypical protein kinase C lambda/iota (aPKCλ) is associated with cell proliferation through unknown mechanisms. In endothelial cells, suppression of
Farivash Darabi et al.
Journal of inorganic biochemistry, 149, 102-107 (2015-04-14)
Aurothiomalate (AuTm) is an old, clinically established, antiarthritic gold drug that is currently being reconsidered as a candidate drug for cancer treatment and for other therapeutic indications within a more general drug repositioning program. As the biological effects of gold

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service