Skip to Content
Merck
All Photos(1)

Key Documents

15512

Sigma-Aldrich

Boc-Trp-OH

≥99.0% (TLC)

Synonym(s):

Nα-(tert-Butoxycarbonyl)-L-tryptophan, Nα-Boc-L-tryptophan

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$105.00

$105.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$105.00

About This Item

Empirical Formula (Hill Notation):
C16H20N2O4
CAS Number:
Molecular Weight:
304.34
Beilstein:
39677
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

$105.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99.0% (TLC)

optical activity

[α]20/D −20±1°, c = 1% in DMF

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

136 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C16H20N2O4/c1-16(2,3)22-15(21)18-13(14(19)20)8-10-9-17-12-7-5-4-6-11(10)12/h4-7,9,13,17H,8H2,1-3H3,(H,18,21)(H,19,20)/t13-/m0/s1

InChI key

NFVNYBJCJGKVQK-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peroxidase catalyzed nitration of tryptophan derivatives
Sala A, et al.
European Journal of Biochemistry, 271(13), 2841-2852 (2004)
Bailey T , et al. et al.
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 21 (2014)
Y Kato et al.
Biochemical and biophysical research communications, 234(1), 82-84 (1997-05-08)
The aim of this study was to clarify the mechanism of loss of Trp residues in proteins exposed to peroxynitrite. The Trp residues in bovine serum albumin and collagen IV were decreased by peroxynitrite treatment. To identify the degradation products
R Magous et al.
Biochimica et biophysica acta, 845(2), 158-162 (1985-05-30)
Benzotript (N-p-chlorobenzoyl-L-tryptophan) has been shown to be a receptor-antagonist in vivo and in vitro for peptides from the gastrin family. In the present study, we examine tryptophan, and some of its N- and C-acylated derivatives, as well as some phenylalanine
R Vespalec et al.
Electrophoresis, 19(2), 276-281 (1998-04-21)
An equation for the calculation of electrophoretic mobility of kinetically labile complexes originating in solutions during the chiral discrimination process is derived. The mobility of the complex is calculated from that of a fully ionized racemic compound, measured in absence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service