Skip to Content
Merck
All Photos(1)

Key Documents

15442

Sigma-Aldrich

Boc-Ile-OSu

≥97.0% (HPLC)

Synonym(s):

Boc-L-isoleucine hydroxysuccinimide ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O6
CAS Number:
Molecular Weight:
328.36
Beilstein:
1552969
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97.0% (HPLC)

form

solid

optical activity

[α]20/D −27±2°, c = 2% in dioxane

mp

103-108 °C

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O

InChI

1S/C15H24N2O6/c1-6-9(2)12(16-14(21)22-15(3,4)5)13(20)23-17-10(18)7-8-11(17)19/h9,12H,6-8H2,1-5H3,(H,16,21)/t9-,12-/m0/s1

InChI key

FATJLEZSGFVHQA-CABZTGNLSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiara Orlandi et al.
Structure (London, England : 1993), 28(5), 516-527 (2020-03-27)
Antibody structure couples adaptive and innate immunity via Fab (antigen binding) and Fc (effector) domains that are connected by unique hinge regions. Because antibodies harbor two or more Fab domains, they are capable of crosslinking multi-determinant antigens, which is required
Xiaofeng Chen et al.
BMC microbiology, 20(1), 264-264 (2020-08-25)
Biofilm is known to be tolerant towards antibiotics and difficult to eradicate. Numerous studies have reported minimum biofilm eradication concentration (MBEC) values of antibiotics for many known biofilm pathogens. However, the experimental parameters applied in these studies differ considerably, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service