Skip to Content
Merck
All Photos(1)

Key Documents

154253

Sigma-Aldrich

4-Bromo-2-methylaniline

97%

Synonym(s):

2-Amino-5-bromotoluene, 4-Bromo-o-toluidine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$95.20

$95.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$95.20

About This Item

Linear Formula:
BrC6H3(CH3)NH2
CAS Number:
Molecular Weight:
186.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$95.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

bp

240 °C (lit.)

mp

57-59 °C (lit.)

functional group

bromo

SMILES string

Cc1cc(Br)ccc1N

InChI

1S/C7H8BrN/c1-5-4-6(8)2-3-7(5)9/h2-4H,9H2,1H3

InChI key

PCHYYOCUCGCSBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromo-2-methylaniline was used in the preparation of 4-Bromo-2-methylphenol[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

QSAR for the cytotoxicity of 2-alkyl or 2, 6-dialkyl, 4-X-phenols: the nature of the radical reaction.
J. Chem. Soc. Perkin Trans. II, 6, 1112-1117 (2002)
Anton Georgiev et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 175, 76-91 (2016-12-27)
In this paper three different "push-pull" 4-aminoazobenzene dyes have been synthesized in order to characterize their photochromic behavior in different solvents. The molecular geometry was optimized by DFT/B3LYP functional combined with the standard 6-31+G(d,p) basis set for trans (E) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service