Skip to Content
Merck
All Photos(1)

Key Documents

152730

Sigma-Aldrich

2-Chloro-6-methoxypyridine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$141.00

$141.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$141.00

About This Item

Empirical Formula (Hill Notation):
C6H6ClNO
CAS Number:
Molecular Weight:
143.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$141.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.528 (lit.)

bp

185-186 °C (lit.)

density

1.207 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

COc1cccc(Cl)n1

InChI

1S/C6H6ClNO/c1-9-6-4-2-3-5(7)8-6/h2-4H,1H3

InChI key

VAVGOGHLNAJECD-UHFFFAOYSA-N

General description

The FT-Raman and Fourier transform infrared (FTIR) spectra of 2-Chloro-6-methoxypyridine has been studied[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Arjunan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(5), 1625-1632 (2011-03-09)
The Fourier transform infrared (FTIR) and FT-Raman spectra of 2-chloro-6-methoxypyridine have been recorded in the range 3700-400 and 3700-100 cm(-1), respectively. The complete vibrational assignment and analysis of the fundamental modes of the compound was carried out using the observed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service