Skip to Content
Merck
All Photos(1)

Key Documents

15033

Sigma-Aldrich

1,3-Di-Boc-2-(trifluoromethylsulfonyl)guanidine

≥95.0% (HPLC)

Synonym(s):

N,N′-Bis(tert-butoxycarboyl)-N′′-trifylguanidine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$156.00
5 G
$669.00

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$156.00
5 G
$669.00

About This Item

Linear Formula:
CF3SO2N=C[NHCO2C(CH3)3]2
CAS Number:
Molecular Weight:
391.36
Beilstein:
7947176
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95.0% (HPLC)

form

solid

mp

113 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N\C(NC(=O)OC(C)(C)C)=N\S(=O)(=O)C(F)(F)F

InChI

1S/C12H20F3N3O6S/c1-10(2,3)23-8(19)16-7(17-9(20)24-11(4,5)6)18-25(21,22)12(13,14)15/h1-6H3,(H2,16,17,18,19,20)

InChI key

GOQZIPJCBUYLIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Guanidinylation reagents for amines and peptides[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdelkader A Metwally et al.
Pharmaceutics, 3(3), 406-424 (2011-01-01)
Four guanidine derivatives of N4,N9-diacylated spermine have been designed, synthesized, and characterized. These guanidine-containing cationic lipids bound siRNA and formed nanoparticles. Two cationic lipids with C18 unsaturated chains, N1,N12-diamidino-N4,N9-dioleoylspermine and N1,N12-diamidino-N4-linoleoyl-N9-oleoylspermine, were more efficient in terms of GFP expression reduction
Tom S Carter et al.
Angewandte Chemie (International ed. in English), 55(32), 9311-9315 (2016-06-18)
Biomimetic carbohydrate receptors ("synthetic lectins") have potential as agents for biological research and medicine. However, although effective strategies are available for "all-equatorial" carbohydrates (glucose, etc.), the recognition of other types of saccharide under natural (aqueous) conditions is less well developed.
K. Feichtinger et al.
The Journal of Organic Chemistry, 63, 8432-8432 (1998)
Maryam Pourhajibagher et al.
Photodiagnosis and photodynamic therapy, 31, 101834-101834 (2020-05-29)
Antimicrobial photodynamic therapy (aPDT) is a treatment to deal with microorganisms, which is limited to treating microbial biofilms due to poor light penetration. Sonodynamic antimicrobial chemotherapy (SACT) can be used for circumventing the limitations of aPDT to inhibit the polymicrobial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service