Skip to Content
Merck
All Photos(1)

Key Documents

15017

Sigma-Aldrich

Boc-Phe(4-CF3)-OH

≥98.0% (TLC)

Synonym(s):

Boc-4-(trifluoromethyl)-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18F3NO4
CAS Number:
Molecular Weight:
333.30
Beilstein:
5617499
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (TLC)

optical activity

[α]20/D +5.4±0.7°, c = 1% in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](Cc1ccc(cc1)C(F)(F)F)C(O)=O

InChI

1S/C15H18F3NO4/c1-14(2,3)23-13(22)19-11(12(20)21)8-9-4-6-10(7-5-9)15(16,17)18/h4-7,11H,8H2,1-3H3,(H,19,22)(H,20,21)/t11-/m0/s1

InChI key

SMVCCWNHCHCWAZ-NSHDSACASA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service