Skip to Content
Merck
All Photos(1)

Key Documents

148229

Sigma-Aldrich

Thiobenzamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CSNH2
CAS Number:
Molecular Weight:
137.20
Beilstein:
606021
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

113-117 °C (lit.)

functional group

amine
phenyl

SMILES string

NC(=S)c1ccccc1

InChI

1S/C7H7NS/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H2,8,9)

InChI key

QIOZLISABUUKJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thiobenzamide was used to prepare amide and amidine adducts[1]. It was also used in the synthesis of 4-oxo-4H-chromene-3-carbothioic acid N-phenylamides[2].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yakov M Koen et al.
Chemical research in toxicology, 26(4), 564-574 (2013-03-08)
Thioacetamide (TA) has long been known as a hepatotoxicant whose bioactivation requires S-oxidation to thioacetamide S-oxide (TASO) and then to the very reactive S,S-dioxide (TASO2). The latter can tautomerize to form acylating species capable of covalently modifying cellular nucleophiles including
D R Doerge et al.
Biochemistry, 30(37), 8960-8964 (1991-09-17)
The mechanism of organosulfur oxygenation by peroxidases [lactoperoxidase (LPX), chloroperoxidase, thyroid peroxidase, and horseradish peroxidase] and hydrogen peroxide was investigated by use of para-substituted thiobenzamides and thioanisoles. The rate constants for thiobenzamide oxygenation by LPX/H2O2 were found to correlate with
J M Domagala et al.
Drug design and discovery, 15(1), 49-61 (1997-05-01)
Substituted 2,2'-dithiobisbenzamides and 2-benzisothiazolones were prepared and shown to possess low microM activity with high therapeutic indices against HIV-1, HIV-2 and SIV in cell culture. The mechanism of antiviral action was determined to be directed toward the nucleocapsid protein (NCp7)
M M Simile et al.
Carcinogenesis, 17(7), 1533-1537 (1996-07-01)
S-Adenosyl-L-methionine (SAM) is a strong chemopreventive agent of rat liver carcinogenesis. Examination was made to determine whether inhibition by SAM of the development of preneoplastic liver lesions persists to SAM withdrawal in diethylnitrosamine-initiated F344 rats promoted with thiobenzamide (TB). The
M Saracoglu et al.
Journal of biomedicine & biotechnology, 2010, doi:10-doi:10 (2010-09-28)
The structure anti-influenza activity relationships of thiobenzamide and quinolizidine derivatives, being influenza fusion inhibitors, have been investigated using the electronic-topological method (ETM) and artificial neural network (ANN) method. Molecular fragments specific for active compounds and breaks of activity were calculated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service