Skip to Content
Merck
All Photos(1)

Key Documents

14804

Sigma-Aldrich

1,2-Bis(dimethylsilyl)benzene

≥98.0%

Synonym(s):

o-Phenylenebis(dimethylsilane)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4[SiH(CH3)2]2
CAS Number:
Molecular Weight:
194.42
Beilstein:
2937027
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

refractive index

n20/D 1.51 (lit.)
n20/D 1.51

bp

101 °C/13 mmHg (lit.)

density

0.898 g/mL at 25 °C (lit.)

SMILES string

C[SiH](C)c1ccccc1[SiH](C)C

InChI

1S/C10H18Si2/c1-11(2)9-7-5-6-8-10(9)12(3)4/h5-8,11-12H,1-4H3

InChI key

QAUCEYVYCBYVDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Protecting group-reagent for amines and amino acids forming "benzostabase" derivatives, more acid-stable than "stabase" analogs; Reaction with olefins and ynes

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F. Cavelier-Frontin et al.
Tetrahedron, 47, 9807-9807 (1991)
M. Tanaka et al.
Organometallics, 10, 16-16 (1991)
R.P. Bonar-Law et al
Tetrahedron Letters, 31, 6721-6721 (1990)
Tetrahedron, 47, 9807-9807 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service