Skip to Content
Merck
All Photos(1)

Key Documents

147508

Sigma-Aldrich

Fertilysin™

97%

Synonym(s):

N,N′-Bis(dichloroacetyl)-1,8-octamethylenediamine, N,N′-Octamethylenebis(2,2-dichloroacetamide), N,N′-Octamethylenebis(dichloroacetamide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20Cl4N2O2
CAS Number:
Molecular Weight:
366.11
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

97%

SMILES string

ClC(Cl)C(=O)NCCCCCCCCNC(=O)C(Cl)Cl

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Momma et al.
Fetal diagnosis and therapy, 9(1), 44-52 (1994-01-01)
Interrupted aortic arch type B associated with ventricular septal defect (n = 17) or atrioventricular septal defect (n = 2) was induced in 19 of 300 fetuses by maternal administration of bis-diamine on the 9th and 10th day of pregnancy.
John K Amory et al.
Journal of andrology, 32(1), 111-119 (2010-08-14)
The bisdichloroacetyldiamine WIN 18,446 reversibly inhibits spermatogenesis in many species, including humans; however, the mechanism by which WIN 18,446 functions is unknown. As retinoic acid is essential for spermatogenesis, we hypothesized that WIN 18,446 might inhibit retinoic acid biosynthesis from
H Tasaka et al.
Teratology, 43(3), 191-200 (1991-03-01)
Administration of N,N'-bis(dichloroacetyl)-1,8-octamethylenediamine, bisdiamine, in pregnant Donryu rats on day 10 of gestation induces a high incidence of cardiovascular anomalies in fetuses. Bisdiamine administration induced aplasia of the sixth aortic arch artery, with both the right and left primitive pulmonary
M Jackson et al.
Cardiovascular research, 30(6), 992-1000 (1995-12-01)
To examine morphological similarities between "classically different" congenital heart lesion types induced in fetal rats by the cardiac teratogen N,N'-bis(dichloroacetyl)-1,8-octamethylenediamine (bis-diamine). A single 200 mg oral dose of bis-diamine was given to 50 pregnant rats on the 9th gestational day.
Takashi Hanato et al.
Birth defects research. Part B, Developmental and reproductive toxicology, 92(1), 10-16 (2011-02-12)
Conotruncal anomalies are often associated with abnormal coronary arteries. Although bis-diamine is known to induce conotruncal defects, its pathological effects on coronary vascular development have not been demonstrated. This study sought to assess the teratogenic effects of bis-diamine on coronary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service