Skip to Content
Merck
All Photos(2)

Key Documents

145947

Sigma-Aldrich

Tetrachlorocyclopropene

98%

Synonym(s):

1,2,3,3-Tetrachlorocyclopropene, Perchlorocyclopropene, Tetrachlorocyclopropenone, Tetrachlorocycloproprene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$485.00
25 G
$1,620.00

$485.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$485.00
25 G
$1,620.00

About This Item

Empirical Formula (Hill Notation):
C3Cl4
CAS Number:
Molecular Weight:
177.84
Beilstein:
1099206
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$485.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.507 (lit.)

bp

125-130 °C (lit.)

density

1.45 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)C1(Cl)Cl

InChI

1S/C3Cl4/c4-1-2(5)3(1,6)7

InChI key

BLZOHTXDDOAASQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrachlorocyclopropene undergoes electrochemical reductive tert-butyldimethylsilylation to yield 1,2,3-tris(tert-butyldimethylsilyl)cyclopropene[1]. It undergoes alkylation reaction with ferrocene in dichloromethane and AlCl3 to yield 2,3-diferrocenylcyclopropenone[2].

Application

Tetrachlorocyclopropene was used to prepare starting reagent for synthesis of 3,3-diethyl-and 3,3-dibenzyl-1,2-diferrocenylcyclopropenes[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Elena I Klimova et al.
Organic & biomolecular chemistry, 1(24), 4458-4464 (2004-01-20)
Reactions of 2,3-diferrocenylcyclopropenone 1 with ethyl- and benzylmagnesium chlorides afford 3,3-diethyl-and 3,3-dibenzyl- 1,2-diferrocenylcyclopropenes 2 and 3, respectively, and products of nucleophilic opening of the three-membered ring resulting from the addition of RMgCl to the carbonyl group, viz., saturated ketones(4,5-diferrocenylheptan-3-ones 4a,b
H A Buchholz et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(18), 10003-10005 (1999-09-01)
Electrochemical reductive tert-butyldimethylsilylation of tetrachlorocyclopropene to 1,2,3-tris(tert-butyldimethylsilyl)cyclopropene, a potential strained precursor for Diels-Alder and related cycloaddition reactions, is described. By hydride abstraction with nitrosonium tetrafluoroborate, 1,2,3-tris(tert-butyldimethylsilyl)cyclopropene is ionized quantitatively to Hückeloid 2pi aromatic tris(tert-butyldimethylsilyl)cyclopropenium tetrafluoroborate.
Noam Y Steinman et al.
Pharmaceutics, 12(8) (2020-08-23)
Non-viral vectors for the transfection of genetic material are at the frontier of medical science. In this article, we introduce for the first time, cyclopropenium-containing nanoparticles as a cationic carrier for gene transfection, as an alternative to the common quaternary
Jesús García-Valdés et al.
Journal of inorganic biochemistry, 197, 110689-110689 (2019-05-18)
Bis-cations with two 2,3-diferrocenylcyclopropenium fragments 3a-d, and the cis-2-(1,2-diferrocenylvinyl)-2-imidazolinium tetrafluoroborates 4a, d or the cis-2-(1,2-diferrocenylvinyl)-3,4,5,6-tetrahydropyrimidin-2-ium tetrafluoroborates 4b, c were obtained by interactions of 2,3-diferrocenyl-1-ethoxycyclopropenium tetrafluoroborate 1 with bis-1,4-N,N-(2a, d) or bis-1,5-N,N-(2b, c) nucleophiles. The reactions of 3a-d with sodium azide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service