Skip to Content
Merck
All Photos(3)

Key Documents

145785

Sigma-Aldrich

1-Bromodecane

98%

Synonym(s):

Decyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$42.80
500 G
$81.20
2 KG
$288.00

$42.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$42.80
500 G
$81.20
2 KG
$288.00

About This Item

Linear Formula:
CH3(CH2)9Br
CAS Number:
Molecular Weight:
221.18
Beilstein:
1735227
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$42.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.456 (lit.)

bp

238 °C (lit.)

density

1.066 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCBr

InChI

1S/C10H21Br/c1-2-3-4-5-6-7-8-9-10-11/h2-10H2,1H3

InChI key

MYMSJFSOOQERIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1-Bromodecane participates in alkylation of pentaerythritol and introduces two lipophilic groups in pentaerythritol[1]. It reacts with 1,2-dimethylimidazole to yield 1-decyl-2,3-dimethylimidazolium bromide[2].

Application

1-Bromodecane was used in the synthesis of ferrocene containing hexacatenar metallomesogen[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Omar F Hernández-Zepeda et al.
Frontiers in plant science, 9, 1076-1076 (2018-08-09)
Plants in more than 100 families secrete extrafloral nectar (EFN) to establish food-for-protection mutualisms with ants. Facultative ant-plants secrete EFN as a jasmonic acid (JA)-dependent response to attract generalist ants. In contrast, obligate ant-plants like the Central American "Swollen-Thorn Acacias"
Isao Kanesaka
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(1-2), 297-302 (2003-12-13)
A change in an infrared intensity in dielectric media is treated by an electrostatic model. The basic model is originally formalized for a dipolar liquid. The model is satisfactorily applied to the infrared intensity of the C-H stretching of chloroform
1,1'-Disubstituted ferrocene containinghexacatenar thermotropic liquid crystals.
Seo JS, et al.
Journal of Materials Chemistry, 11(5), 1332-1338 (2001)
Xuejing Song et al.
Materials (Basel, Switzerland), 11(10) (2018-10-27)
Pyrazine derivatives quinoxaline and pyridopyrazine were selected as the acceptors, and benzocarbazole was used as the donor to synthesize four different D⁻A⁻D compounds. The results showed that 2,3-bis(decyloxy)pyridine[3,4-b]pyrazine (DPP) exhibited stronger electron-withdrawing ability than that of 2,3-bis(decyloxy)quinoxaline (DPx), because DPP

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service