Skip to Content
Merck
All Photos(1)

Key Documents

145300

Sigma-Aldrich

3,4-(Methylenedioxy)phenylacetonitrile

99%

Synonym(s):

1,3-Benzodioxole-5-acetonitrile, Homopiperonylnitrile

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$138.00

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$138.00

About This Item

Empirical Formula (Hill Notation):
C9H7NO2
CAS Number:
Molecular Weight:
161.16
Beilstein:
7739
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

mp

43-45 °C (lit.)

functional group

nitrile

SMILES string

N#CCc1ccc2OCOc2c1

InChI

1S/C9H7NO2/c10-4-3-7-1-2-8-9(5-7)12-6-11-8/h1-2,5H,3,6H2

InChI key

ZQPBOYASBNAXOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-(Methylenedioxy)phenylacetonitrile undergoes hydrolysis catalyzed by nitrilase ZmNIT2 enzyme from maize[1].

Application

3,4-(Methylenedioxy)phenylacetonitrile was used in synthesis of derrubone[2]. It was used as standard to analyze the seized methamphetamine samples showing unique profiles of stable isotopic compositions by isotope ratio mass spectrometry[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seized methamphetamine samples with unique profiles of stable nitrogen isotopic composition documented by stable isotope ratio mass spectrometry.
Iwata YT, et al.
Forensic Toxicology, 28(2), 119-123 (2010)
Jedidiah M Hastings et al.
The Journal of organic chemistry, 73(2), 369-373 (2007-12-25)
Recently, we reported that the natural product derrubone exhibits Hsp90 inhibitory activity. Due to its unique architectural scaffold and proposed rapid assembly, the synthesis of this natural product was pursued with the aim of identifying structure--activity relationships. Synthesis of the
Enzymatic nitrile hydrolysis catalyzed by nitrilase ZmNIT2 from maize. An unprecedented ?-hydroxy functionality enhanced amide formation.
Mukherjee C, et al.
Tetrahedron, 62(26), 6150-6154 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service