Skip to Content
Merck
All Photos(1)

Key Documents

145009

Sigma-Aldrich

2,4-Dichlorotoluene

99%

Synonym(s):

1,3-Dichloro-4-methylbenzene, 2,4-Dichloro-1-methylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$76.30
1 KG
$256.00

$76.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$76.30
1 KG
$256.00

About This Item

Linear Formula:
CH3C6H3Cl2
CAS Number:
Molecular Weight:
161.03
Beilstein:
1931691
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$76.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.546 (lit.)

bp

200 °C (lit.)

density

1.246 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Cc1ccc(Cl)cc1Cl

InChI

1S/C7H6Cl2/c1-5-2-3-6(8)4-7(5)9/h2-4H,1H3

InChI key

FUNUTBJJKQIVSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2,4-Dichlorotoluene was used as an oxidant and a solvent during anaerobic catalytic oxidation of secondary alcohols by using an in situ (N-heterocyclic carbene)-Ni(0) system[1]. It was used as growth supplement in the culture media of Ralstonia sp. strain PS12[2]. It was used to develop a sensitive method based on solid-phase microextraction followed by gas chromatography-tandem mass spectrometry for the analysis of chlorotoluenes in water samples[3].

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Christophe Berini et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(23), 6857-6860 (2010-05-11)
The selective, anaerobic catalytic oxidation of secondary alcohols at room temperature by using an in situ (N-heterocyclic carbene)-Ni(0) system is presented. The use of non-anhydrous, non-degassed 2,4-dichlorotoluene as both the oxidant and the solvent allows for very short reaction times
V Karunakaran et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 95, 64-72 (2012-05-23)
The FT-IR and FT-Raman spectra of α-bromo-2,6-dichlorotoluene (αBDCT) have been recorded. The structural and spectroscopic data of the molecule in the ground state have been calculated using Hartree Fock (HF) and Density Functional Theory (DFT)/B3LYP with the standard 6-31++G(d,p) basis
[Hygienic standards for 2,4-dichlorotoluene in reservoir water].
G V Tolstpiatova et al.
Gigiena i sanitariia, (2)(2), 80-81 (1988-02-01)
Katrin Pollmann et al.
Journal of bacteriology, 184(19), 5261-5274 (2002-09-10)
Ralstonia sp. strain PS12 is able to use 2,4-, 2,5-, and 3,4-dichlorotoluene as growth substrates. Dichloromethylcatechols are central intermediates that are formed by TecA tetrachlorobenzene dioxygenase-mediated activation at two adjacent unsubstituted carbon atoms followed by TecB chlorobenzene dihydrodiol dehydrogenase-catalyzed rearomatization
[Gas chromatographic determination of 2,4-dichlorotoluene in water].
T A Kozeĭko et al.
Gigiena i sanitariia, (3)(3), 44-45 (1988-03-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service