Skip to Content
Merck
All Photos(1)

Documents

144274

Sigma-Aldrich

2,5-Dichlorobenzenethiol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3SH
CAS Number:
Molecular Weight:
179.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

112-116 °C/50 mmHg (lit.)

SMILES string

Sc1cc(Cl)ccc1Cl

InChI

1S/C6H4Cl2S/c7-4-1-2-5(8)6(9)3-4/h1-3,9H

InChI key

QIULLHZMZMGGFH-UHFFFAOYSA-N

Application

2,5-Dichlorobenzenethiol may be used in the synthesis of radiolabeled alkyl aryl ethers and sulphides via phase transfer catalysis and chlorinated biphenyl methylsulfonyl metabolites.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An alternative synthesis of chlorinated biphenyl methylsulfonyl metabolites.
Mortimer RD and Newsome WH.
Chemosphere, 32(5), 935-946 (1996)
Phase transfer mediated synthesis of radiolabelled alkyl aryl ethers and sulphides.
Bergman A and Wachtmeister CA.
Journal of Labelled Compounds & Radiopharmaceuticals, 24(8), 925-930 (1987)
Esma Khatun et al.
Nanoscale, 9(24), 8240-8248 (2017-06-06)
We report the synthesis of a new silver cluster, [Ag

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service