Skip to Content
Merck
All Photos(2)

Key Documents

143979

Sigma-Aldrich

Tripropylamine

≥98%

Synonym(s):

N,N,N-Tri-n-propylamine, N,N-Dipropyl-1-propanamine, Propyldi-n-propylamine, Tri-n-propylamine

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$32.10
500 ML
$75.20
2 L
$208.00
18 L
$578.00

$32.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$32.10
500 ML
$75.20
2 L
$208.00
18 L
$578.00

About This Item

Linear Formula:
(CH3CH2CH2)3N
CAS Number:
Molecular Weight:
143.27
Beilstein:
1098331
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$32.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

4.9 (vs air)

Quality Level

vapor pressure

2.9 mmHg ( 20 °C)

Assay

≥98%

form

liquid

autoignition temp.

356 °F

expl. lim.

5.6 %

refractive index

n20/D 1.416 (lit.)

bp

155-158 °C (lit.)
40-42 °C/11 mmHg (lit.)

mp

−93.5 °C (lit.)

density

0.753 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCN(CCC)CCC

InChI

1S/C9H21N/c1-4-7-10(8-5-2)9-6-3/h4-9H2,1-3H3

InChI key

YFTHZRPMJXBUME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Eelectrochemiluminescence behavior of ruthenium complex/tripropylamine systems at DNA-modified gold electrode has been studied[1].

Application

Tripropylamine was employed as coreactant in sandwich-type electrochemiluminescence biosensor for detection of α-fetoprotein[2]. It was used as extraction agent for the reactive extraction of succinic acid in various alcohol diluents[3].

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong-Yuan Liu et al.
Biosensors & bioelectronics, 26(5), 2703-2706 (2010-10-01)
The electrochemiluminescence (ECL) behavior of ruthenium complex/tripropylamine (TPA) systems at DNA-modified gold electrode was studied to understand the possible mechanism and to develop new detection platforms. DNA strand, especially double-stranded DNA (ds-DNA), can preconcentrate TPA and acts as the acceptor
Reactive extraction of succinic acid with tripropylamine (TPA) in various diluents.
Hong YK and Hong WH.
Bioprocess and Biosystems Engineering, 22(4), 281-284 (2000)
Suli Liu et al.
Nanoscale, 6(4), 2419-2425 (2014-01-18)
Using ruthenium polypyridyl functionalized ZnO mesocrystals as bionanolabels, a universal biological recognition and biosensing platform based on gold nanoparticle (AuNP) dotted reduced graphene oxide (rGO) composite was developed. AuNP-rGO accelerated electron transfer between the detection probe and the electrode, and
H C Froede et al.
Archives of biochemistry and biophysics, 247(2), 420-423 (1986-06-01)
The theory of noncompetitive inhibition of acetylcholinesterase based on the binding of inhibitor to the acetylenzyme and the free enzyme was proven correct by demonstrating that tripropylammonium ion increases the steady-state concentration of acetylenzyme, as predicted by the theory. By
Xueshu Li et al.
Langmuir : the ACS journal of surfaces and colloids, 28(9), 4152-4159 (2012-02-07)
The micelles of two tripropargylammonium-functionalized cationic surfactants were cross-linked by a disulfide-containing diazido cross-linker in the presence of Cu(I) catalysts. With multiple residual alkyne groups on the surface, the resulting surface cross-linked micelles (SCMs) were postfunctionalized by reaction with 2-azidoethanol

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service