Skip to Content
Merck
All Photos(1)

Key Documents

140201

Sigma-Aldrich

1-Phenyl-1-cyclopentanecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$62.60

$62.60


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$62.60

About This Item

Linear Formula:
C6H5C5H8CO2H
CAS Number:
Molecular Weight:
190.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$62.60


Estimated to ship onMay 26, 2025


Request a Bulk Order

Assay

98%

form

solid

mp

159-161 °C (lit.)

functional group

carboxylic acid
phenyl

SMILES string

OC(=O)C1(CCCC1)c2ccccc2

InChI

1S/C12H14O2/c13-11(14)12(8-4-5-9-12)10-6-2-1-3-7-10/h1-3,6-7H,4-5,8-9H2,(H,13,14)

InChI key

RHPCYZLXNNRRMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Phenyl-1-cyclopentanecarboxylic acid was used to prepare precursors required for solid phase synthesis of isoxazole and isoxazoline derivatives[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sureshbabu Dadiboyena et al.
Tetrahedron letters, 53(16), 2096-2099 (2012-06-21)
An efficient approach for the parallel solid phase synthesis of isoxazole and isoxazoline derivatives has been developed. The isoxazoles and isoxazolines were constructed through a 1,3-dipolar cycloaddition reaction of nitrile oxides, with resin-bound alkynes or alkenes. The cycloaddition reaction conditions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service