Skip to Content
Merck
All Photos(1)

Key Documents

139432

Sigma-Aldrich

1,3,4-Thiadiazole-2,5-dithiol dipotassium salt

1,3,4-Thiadiazole-2,5-dithiol dipotassium salt

98%

Synonym(s):

Bismuthiol I dipotassium salt, 2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt, Bismuththiol I dipotassium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$134.00

$134.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$134.00

About This Item

Empirical Formula (Hill Notation):
C2K2N2S3
CAS Number:
Molecular Weight:
226.43
Beilstein:
4917786
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$134.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

274-276 °C (dec.) (lit.)

SMILES string

[K]Sc1nnc(S[K])s1

InChI

1S/C2H2N2S3.2K/c5-1-3-4-2(6)7-1;;/h(H,3,5)(H,4,6);;/q;2*+1/p-2

InChI key

GPWLFGDMYSVEGN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

1,3,4-thiadiazole-2,5-dithiol undergoes copolymerization with 1+, 2+ and 3+ valent metals to form coordination polymers[1]. It forms polymers with metals and can be useful in treatment of metal-polluted water[2].

Application

1,3,4-thiadiazole-2,5-dithiol (Bismuthiol I) was used in determination of tellurium in indium phosphide doped with cadmium telluride by electrothermal atomic absorption spectrometry[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of metallic azoderivatives of 2-amino-5-mercapto-1, 3, 4-thiadiazole.
Ortega-Luoni P, et al.
Journal of the Chilean Chemical Society, 52(1), 1120-1122 (2007)
Determination of tellurium in indium phosphide by electrothermal atomic absorption spectrometry and ultraviolet-visible spectrophotometry.
Taddia M, et al.
Journal of Analytical Atomic Spectrometry, 10(6), 433-437 (1995)
Coordination polymers from 1, 3, 4-thiadiazole-2, 5-dithiol and metal ions.
Ortega P, etal.
Macromolecular Chemistry and Physics, 198(9), 2949-2956 (1997)
N Vasimalai et al.
Journal of hazardous materials, 213-214, 193-199 (2012-02-22)
This manuscript describes a highly selective and ultra sensitive determination of Hg(II) in aqueous solution using functionalized mercaptothiadiazole capped silver nanoparticles (AgNPs) by spectrofluorimetry. We have synthesized 2,5-dimercapto-1,3,4-thiadiazole (DMT), 2-mercapto-5-methyl-1,3,4-thiadiazole (MMT) and 2-mercapto-5-amino-1,3,4-thiadiazole (AMT) capped AgNPs by wet chemical method.
W F Herblin et al.
Life sciences, 56(15), 1257-1264 (1995-03-03)
A rapid, high capacity assay for the binding of basic fibroblast growth factor has been developed. Rat lung tissue was selected as the optimum source of membranes and 2M sodium chloride used to remove endogenous growth factor. The assay has

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service