136387
4-(Fluorosulfonyl)benzoyl chloride
technical grade, 90%
Synonym(s):
p-(Fluorosulfonyl)benzoyl chloride
Select a Size
Select a Size
About This Item
Recommended Products
grade
technical grade
Assay
90%
reaction suitability
reaction type: click chemistry
bp
96-97 °C/0.7 mmHg (lit.)
mp
46-48 °C (lit.)
functional group
acyl chloride
SMILES string
FS(=O)(=O)c1ccc(cc1)C(Cl)=O
InChI
1S/C7H4ClFO3S/c8-7(10)5-1-3-6(4-2-5)13(9,11)12/h1-4H
InChI key
JMTAYFNTRRLWQG-UHFFFAOYSA-N
1 of 4
This Item | QBD10501 | QBD10567 | QBD11366 |
---|---|---|---|
reaction suitability reaction type: click chemistry | reaction suitability reaction type: click chemistry, reagent type: cross-linking reagent | reaction suitability reaction type: click chemistry, reagent type: cross-linking reagent | reaction suitability reaction type: click chemistry, reagent type: cross-linking reagent |
Quality Level 100 | Quality Level - | Quality Level - | Quality Level - |
mp 46-48 °C (lit.) | mp - | mp - | mp - |
bp 96-97 °C/0.7 mmHg (lit.) | bp - | bp - | bp - |
grade technical grade | grade - | grade - | grade - |
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service