Skip to Content
Merck
All Photos(1)

Key Documents

136085

Sigma-Aldrich

2-Amino-6-chlorobenzothiazole

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$33.60

$33.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$33.60

About This Item

Empirical Formula (Hill Notation):
C7H5ClN2S
CAS Number:
Molecular Weight:
184.65
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$33.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

mp

199-201 °C (lit.)

functional group

chloro

SMILES string

Nc1nc2ccc(Cl)cc2s1

InChI

1S/C7H5ClN2S/c8-4-1-2-5-6(3-4)11-7(9)10-5/h1-3H,(H2,9,10)

InChI key

VMNXKIDUTPOHPO-UHFFFAOYSA-N

General description

2-amino-6-chloro-benzothiazole on microwave irradiation in the presence of 1-butyl-3-methylimidazolium and inorganic anions yields fluorinated benzothiazolo[2,3-b]quinazoline-2H-ones analogues[1]. It has synergistic effect on inhibitive performance of propargyl alcohol during corrosion of mild steel in boiling HCl solution[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synergistic effect of 2-amino-6-chloro-benzothiazole on inhibitive performance of propargyl alcohol during corrosion of mild steel in boiling hydrochloric acid solution.
Quraishi MA, et al.
Bulletin of Electrochemistry, 13(06), 257-259 (1997)
Greener synthesis and photo-antiproliferative activity of novel fluorinated benzothiazolo [2, 3-b] quinazolines.
Arya K, et al. et al.
Medicinal Chemistry Research, 1-9 null
Jonathan R LaRochelle et al.
Bioorganic & medicinal chemistry, 25(24), 6479-6485 (2017-11-02)
The PTPN11 oncogene encodes the cytoplasmic protein tyrosine phosphatase SHP2, which, through its role in multiple signaling pathways, promotes the progression of hematological malignancies and other cancers. Here, we employ high-throughput screening to discover a lead chemical scaffold, the benzothiazolopyrimidones
Ahmed S M Al-Janabi et al.
Journal of molecular structure, 1228, 129454-129454 (2020-10-27)
New Schiff bases {N'-(phenyl(pyridin-2-yl)methylene) isonicotinohydrazide (L1H), N1 -(naphthalen-1-yl)-N2 -(phenyl(pyridin-2-yl) methylidene) ethane-1,2-diamine (L2H), N-(6-chlorobenzo[d]thiazol-2-yl)-1-phenyl-1-(pyridin-2-yl) methanimine (L3H)}were synthesized by reaction of 2-benzoylpyridine with different amines (2-amino-6-chlorobenzothiazole, isonicotinohydrazide and N 1-(naphthalen-1-yl)ethane-1,2-diamine) and characterized by 1H-NMR, 13C-NMR, IR mass spectroscopy and elemental analysis. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service