Skip to Content
Merck
All Photos(1)

Key Documents

132160

Sigma-Aldrich

3,4,5-Trimethoxybenzonitrile

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$104.00

$104.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$104.00

About This Item

Linear Formula:
(CH3O)3C6H2CN
CAS Number:
Molecular Weight:
193.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$104.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

powder

bp

180-185 °C/10 mmHg (lit.)

mp

91-94 °C (lit.)

SMILES string

COc1cc(cc(OC)c1OC)C#N

InChI

1S/C10H11NO3/c1-12-8-4-7(6-11)5-9(13-2)10(8)14-3/h4-5H,1-3H3

InChI key

OSBQUSPVORCDCU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Room-temperature discotic liquid-crystalline coronene diimides exhibiting high charge-carrier mobility in air.
An Z,et al.
Journal of Materials Chemistry, 19(37), 6688-6698 (2009)
Characterization of nitrile hydratation catalysed by Nocardia sp. 108.
Wang Y-J, et al.
World Journal of Microbiology & Biotechnology, 23(3), 355-362 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service