Skip to Content
Merck
All Photos(1)

Documents

131296

Sigma-Aldrich

Oxalyldihydrazide

98%

Synonym(s):

Oxalic dihydrazide, Oxalylhydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NHCOCONHNH2
CAS Number:
Molecular Weight:
118.09
Beilstein:
1072110
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

242-244 °C (dec.) (lit.)

SMILES string

NNC(=O)C(=O)NN

InChI

1S/C2H6N4O2/c3-5-1(7)2(8)6-4/h3-4H2,(H,5,7)(H,6,8)

InChI key

SWRGUMCEJHQWEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Oxalyldihydrazide was used as a crosslinker for enrichment of carbonylated proteins within a microfluid chip. It was also used to synthesize a new series of manganese and iron salt forming complexes by template condensation with glyoxal.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khlood Abou Melha
Journal of enzyme inhibition and medicinal chemistry, 23(2), 285-295 (2008-03-18)
The Schiff base ligand, oxalyl [( 2 - hydroxybenzylidene) hydrazone] [corrected].H(2)L, and its Cu(II), Ni(II), Co(II), UO(2)(VI) and Fe(III) complexes were prepared and tested as antibacterial agents. The Schiff base acts as a dibasic tetra- or hexadentate ligand with metal
D P Singh et al.
Journal of enzyme inhibition and medicinal chemistry, 24(3), 883-889 (2008-10-29)
A new series of complexes is synthesized by template condensation of oxalyldihydrazide and glyoxal in methanolic medium in the presence of trivalent chromium, manganese and iron salts forming complexes of the type: [M(C(8)H(8)N(8)O(4))X]X(2) where M = Cr(III), Mn(III), Fe(III) and
Bryant C Hollins et al.
Lab on a chip, 12(14), 2526-2532 (2012-05-09)
We report a proof of principle study for the use of oxalyldihydrazide as a crosslinker for enrichment of carbonylated proteins within a microfluidic chip. Surface modification steps are characterized and analyzed using analytical techniques. We use oxidized cytochrome c as
Zahid H Chohan et al.
Journal of enzyme inhibition and medicinal chemistry, 17(1), 1-7 (2002-10-09)
Schiff bases derived from oxaldiamide/oxalylhydrazine and pyrrol-2-carbaldehyde, or salicylaldehyde respectively, as well as their Zn(II) complexes have been prepared and tested as antibacterial agents. These Schiff bases function as tetradentate ligands, forming octahedral Zn(II) complexes. The ketonic form for the
M Rizk et al.
Acta pharmaceutica Hungarica, 62(4), 158-166 (1992-07-01)
A sensitive and selective colorimetric method has been devised for the micro-determination of copper (II) ions in pure form, some chemicals and multivitamin preparations. The method depends on the formation of stable blue complexes in ammonia medium peaking at 615

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service