Skip to Content
Merck
All Photos(3)

Key Documents

131202

Sigma-Aldrich

Dihexylamine

97%

Synonym(s):

Bis(1-hexyl)amine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$132.00
500 ML
$437.00

$132.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$132.00
500 ML
$437.00

About This Item

Linear Formula:
[CH3(CH2)5]2NH
CAS Number:
Molecular Weight:
185.35
Beilstein:
1738519
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$132.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

6.4 (vs air)

Assay

97%

form

liquid

refractive index

n20/D 1.432 (lit.)

bp

192-195 °C (lit.)

density

0.795 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCCCCNCCCCCC

InChI

1S/C12H27N/c1-3-5-7-9-11-13-12-10-8-6-4-2/h13H,3-12H2,1-2H3

InChI key

PXSXRABJBXYMFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dihexylamine nitrosation rate was enhanced when the reaction was carried out in the presence of micelles containing bile acid conjugates and lecithin[1].

Application

Dihexylamine was used in the preparation of succinic acid[2].

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y K Kim et al.
IARC scientific publications, (31)(31), 207-214 (1980-01-01)
The rate of nitrosation of dihexylamine is enhanced when the reaction is carried ut in the presence of micelles containing bile acid conjugates or bile acid conjugates and lecithin. The nitrosation of smaller dialkylamines is not affected. The nitrosation of
Tanja Kurzrock et al.
Bioprocess and biosystems engineering, 34(7), 779-787 (2011-02-26)
Biotechnologically produced succinic acid has the potential to displace maleic acid and its uses and to become an important feedstock for the chemical industry. In addition to optimized production strains and fermentation processes, an efficient separation of succinic acid from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service