Skip to Content
Merck
All Photos(1)

Key Documents

130877

Sigma-Aldrich

5-Aminoindan

95%

Synonym(s):

5-Indanamine

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$142.00

$142.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$142.00

About This Item

Empirical Formula (Hill Notation):
C9H11N
CAS Number:
Molecular Weight:
133.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$142.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

95%

form

solid

bp

247-249 °C/745 mmHg (lit.)

mp

34-36 °C (lit.)

SMILES string

Nc1ccc2CCCc2c1

InChI

1S/C9H11N/c10-9-5-4-7-2-1-3-8(7)6-9/h4-6H,1-3,10H2

InChI key

LEWZOBYWGWKNCK-UHFFFAOYSA-N

Application

5-Aminoindan was used in the synthesis of ligand 18/18 (2-(5-aminoindan)-6-(5-aminoindan)-4-chloro-s-triazine)[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U D Palanisamy et al.
Journal of chromatography. B, Biomedical sciences and applications, 746(2), 265-281 (2000-11-15)
A combination of rational design based on mimicking natural protein-carbohydrate interactions and solid-phase combinatorial chemistry has led to the identification of an affinity ligand which displays selectivity for the mannose moiety of glycoproteins. The ligand, denoted 18/18 and comprising a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service