Skip to Content
Merck
All Photos(1)

Key Documents

130613

Sigma-Aldrich

5-Bromoveratraldehyde

98%

Synonym(s):

3-Bromo-4,5-dimethoxybenzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$163.00

$163.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$163.00

About This Item

Linear Formula:
BrC6H2(OCH3)2CHO
CAS Number:
Molecular Weight:
245.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$163.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

98%

form

powder

mp

63-64 °C (lit.)

functional group

aldehyde
bromo

SMILES string

COc1cc(C=O)cc(Br)c1OC

InChI

1S/C9H9BrO3/c1-12-8-4-6(5-11)3-7(10)9(8)13-2/h3-5H,1-2H3

InChI key

ICVODPFGWCUVJC-UHFFFAOYSA-N

Application

5-Bromoveratraldehyde (3-Bromo-4,5-dimethoxybenzaldehyde) was used to prepare 1-phenyl-2-(4-aryl-1,3,4,5-tetrahydropyrido[2,3-b][1,4]diazepin-2-ylidene)-ethanones[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Liszkiewicz et al.
Die Pharmazie, 61(6), 517-521 (2006-07-11)
A two-step, general synthesis of 1-phenyl-2-(4-aryl-1,3,4,5-tetrahydropyrido[2,3-b][1,4]diazepin-2-ylidene)-ethanones 3-9 is presented. This synthesis employs a condensation of 2,3-diaminopyridine with benzoylacetone followed by a basic-activated cyclization reaction with substituted benzaldehydes for final closure of the seven-membered ring. Molecular diversity is fixed by appropriate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service