Skip to Content
Merck
All Photos(1)

Key Documents

129208

Sigma-Aldrich

4-Methoxytriphenylmethyl chloride

97%

Synonym(s):

4-Methoxytriphenylchloromethane, 4-Methoxytrityl chloride, p-Anisylchlorodiphenylmethane, p-Monomethoxytrityl chloride, MMT

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$68.70
25 G
$181.00
100 G
$648.00

$68.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$68.70
25 G
$181.00
100 G
$648.00

About This Item

Linear Formula:
CH3OC6H4C(C6H5)2Cl
CAS Number:
Molecular Weight:
308.80
Beilstein:
798425
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$68.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

mp

122-124 °C (lit.)

functional group

chloro
phenyl

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(Cl)(c2ccccc2)c3ccccc3

InChI

1S/C20H17ClO/c1-22-19-14-12-18(13-15-19)20(21,16-8-4-2-5-9-16)17-10-6-3-7-11-17/h2-15H,1H3

InChI key

OBOHMJWDFPBPKD-UHFFFAOYSA-N

Application

Selective protecting reagent for primary hydroxyl groups.[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Communications, 23, 1239-1239 (1993)
A Wissner et al.
Journal of medicinal chemistry, 35(9), 1650-1662 (1992-05-01)
A series of aryl phosphoglyceride (3, 19-61) and bis-aryl phosphate (67-135) antagonists of platelet activating factor (PAF) were prepared. A group of four bifunctional phosphorus reagents (5a-c and 7) were developed that allowed the preparation of these aryl phosphates in
Dongfen Yuan et al.
Journal of controlled release : official journal of the Controlled Release Society, 263, 172-184 (2017-03-28)
Leptin is an adipocyte-secreted hormone that is delivered via a specific transport system across the blood-brain barrier (BBB) to the brain where it acts on the hypothalamus receptors to control appetite and thermogenesis. Peripheral resistance to leptin due to its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service