Skip to Content
Merck
All Photos(3)

Key Documents

128279

Sigma-Aldrich

Ethyl hydrazinoacetate hydrochloride

97%

Synonym(s):

(Carbethoxymethyl)hydrazine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$70.20
25 G
$233.00

$70.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$70.20
25 G
$233.00

About This Item

Linear Formula:
H2NNHCH2CO2C2H5 · HCl
CAS Number:
Molecular Weight:
154.60
Beilstein:
3912112
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$70.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

powder

mp

152-154 °C (lit.)

functional group

amine
ester
hydrazine

SMILES string

Cl[H].CCOC(=O)CNN

InChI

1S/C4H10N2O2.ClH/c1-2-8-4(7)3-6-5;/h6H,2-3,5H2,1H3;1H

InChI key

HZZRIIPYFPIKHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Ethyl hydrazinoacetate hydrochloride has been used in the preparation of three novel copper(II) complexes as condensation derivatives of 2-pyridinecarboxaldehyde, 2-acetylpyridine and 2-quinolinecarboxaldehyde[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper (II) complexes of N heteroaromatic hydrazones: Synthesis, X-ray structure, magnetic behavior, and antibacterial activity.
Inorgorganica Chimica Acta, 362(6), 1996-2000 (2009)
Nenad Filipović et al.
Chemical biology & drug design, 84(3), 333-341 (2014-03-19)
Novel Pd(II) complex with N-heteroaromatic Schiff base ligand, derived from 8-quinolinecarboxaldehyde (q8a) and ethyl hydrazinoacetate (haOEt), was synthesized and characterized by analytical and spectroscopy methods. The structure of novel complex, as well as structures of its quinoline and pyridine analogues

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service