Skip to Content
Merck
All Photos(2)

Key Documents

127108

Sigma-Aldrich

N,N,N′,N′-Tetramethyl-1,4-butanediamine

98%

Synonym(s):

1,4-Bis(dimethylamino)butane, N,N,N′,N′-Tetramethyl-1,4-diaminobutane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$129.00

$129.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$129.00

About This Item

Linear Formula:
(CH3)2N(CH2)4N(CH3)2
CAS Number:
Molecular Weight:
144.26
Beilstein:
1735538
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$129.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.428 (lit.)

bp

166-167 °C (lit.)

density

0.792 g/mL at 25 °C (lit.)

SMILES string

CN(C)CCCCN(C)C

InChI

1S/C8H20N2/c1-9(2)7-5-6-8-10(3)4/h5-8H2,1-4H3

InChI key

VEAZEPMQWHPHAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Quemener et al.
Biology of the cell, 77(2), 195-199 (1993-01-01)
In a previous study, we identified regions on the surface of tumor cells which act as acceptor sites for putrescine (Put) and studied the competition between structural analogs of Put (N,N'-tetramethyl-alpha,omega-diaminoalkanes) and Put bound to latex microspheres. A chain of
Jamil S Saad et al.
Inorganic chemistry, 50(10), 4559-4571 (2011-04-23)
The highly distorted Pt(d(G*pG*)) (G* = N7-platinated G) 17-membered macrocyclic ring formed by cisplatin anticancer drug binding to DNA alters the structure of the G*G* base pair steps, canting one base, and increases dynamic motion, complicating solution structural studies. However
Chien-Shun Chen et al.
ChemSusChem, 6(1), 132-140 (2012-09-05)
The recycling of process water from strip mining extractions is a very relevant task both industrially and environmentally. The sedimentation of fine tailings during such processes, however, can often require long periods of time and/or the addition of flocculants which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service