Skip to Content
Merck
All Photos(1)

Documents

12490

Sigma-Aldrich

11H-Benzo[a]fluorene

≥98.0%

Synonym(s):

1,2-Benzofluorene, Chrysofluorene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12
CAS Number:
Molecular Weight:
216.28
Beilstein:
2046363
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

mp

185-190 °C

solubility

dioxane: soluble 0.1 g/10 mL, clear, colorless

SMILES string

C1c2ccccc2-c3ccc4ccccc4c13

InChI

1S/C17H12/c1-3-7-14-12(5-1)9-10-16-15-8-4-2-6-13(15)11-17(14)16/h1-10H,11H2

InChI key

HKMTVMBEALTRRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

11H-Benzo[a]fluorene has been used in a study on 7H-benzo[c]fluorine, major lung DNA adductor component in coal tar.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radoslaw S Laufer et al.
Journal of the American Chemical Society, 124(9), 1854-1855 (2002-02-28)
Theoretical and chemical studies of the reactivity of isoprekinamycin, the kinamycins, and the lomaiviticins support the proposal that these natural products exhibit enhanced diazonium salt character and may owe their antitumor antibiotic properties to their ability to act as electrophilic
A Koganti et al.
Carcinogenesis, 21(8), 1601-1609 (2000-07-27)
Coal tar is a complex mixture that exhibits high carcinogenic potency in lungs of animals when administered in the diet. Studies have noted that lung tumor induction does not correlate with the benzo[a]pyrene content of coal tar, suggesting that other
Benzo[a]fluorene.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 32, 177-182 (1983-12-01)
C Atienza et al.
Organic letters, 3(2), 153-155 (2001-06-30)
[figure: see text] Thermal cyclization of 1-[2-(trimethylsilylethynyl)phenyl]-3-arylpropinones was expected to give benzo[b]fluorenones. However, benzo[a]-fluorenones were also formed as a result of a new rearrangement. These tetracycles possess the core structure of the fluostatins and isoprekinamycin.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service