Skip to Content
Merck
All Photos(1)

Key Documents

124303

Sigma-Aldrich

tert-Butyl phenyl carbonate

98%

Synonym(s):

1,1-Dimethylethyl phenyl carbonate, 1-tert-Butoxycarbonyloxybenzene, Phenyl tert-butyl carbonate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$94.30

$94.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$94.30

About This Item

Linear Formula:
(CH3)3COCOOC6H5
CAS Number:
Molecular Weight:
194.23
Beilstein:
2361602
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$94.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

bp

83 °C/0.6 mmHg (lit.)

density

1.05 g/mL at 20 °C (lit.)

functional group

carbonate
phenoxy

SMILES string

CC(C)(C)OC(=O)Oc1ccccc1

InChI

1S/C11H14O3/c1-11(2,3)14-10(12)13-9-7-5-4-6-8-9/h4-8H,1-3H3

InChI key

UXWVQHXKKOGTSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

tert-Butyl phenyl carbonate was used in determination of octanol-water partition coefficients by microemulsion electrokinetic chromatography[1]. It was also used in the synthesis of 2-nitroindoles[2]. It was used as reagent for mono-Boc protection of α,ω-diamines[3].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2-nitroindoles via the Sundberg indole synthesis.
Pelkey ET and Gribble GW.
Tetrahedron Letters, 38(32), 5603-5606 (1997)
Jesper Østergaard et al.
Electrophoresis, 24(6), 1038-1046 (2003-03-27)
Microemulsion electrokinetic chromatography (MEEKC) was assessed as a tool for determination of octanol-water partition coefficients using 34 solutes encompassing 8 carbonate esters. It was confirmed that microemulsions containing 1.44-2.88% w/w SDS, 6.49% w/w 1-butanol, and 0.82% w/w n-heptane constitute a
Organic Syntheses, 84, 209-209 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service