Skip to Content
Merck
All Photos(4)

Key Documents

123986

Sigma-Aldrich

Bromoacetaldehyde diethyl acetal

97%

Synonym(s):

2-Bromo-1,1-diethoxyethane, Bromoacetal

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$48.10
500 G
$228.00
1 KG
$291.00

$48.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$48.10
500 G
$228.00
1 KG
$291.00

About This Item

Linear Formula:
BrCH2CH(OC2H5)2
CAS Number:
Molecular Weight:
197.07
Beilstein:
635754
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$48.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

66-67 °C/18 mmHg (lit.)

density

1.31 g/mL at 25 °C (lit.)

functional group

bromo
ether

storage temp.

2-8°C

SMILES string

CCOC(CBr)OCC

InChI

1S/C6H13BrO2/c1-3-8-6(5-7)9-4-2/h6H,3-5H2,1-2H3

InChI key

LILXDMFJXYAKMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromoacetaldehyde diethyl acetal reacts with anhydrous trimethylamine to form glycine betaine aldehyde[1]. It reacts with different thiophenols in the presence of KOH/Cu to produce the corresponding ketone and diethyl acetals[2].
May darken in storage

Application

Bromoacetaldehyde diethyl acetal has been used in the synthesis of monomer 2-(2,2-dimethoxy)ethoxyethyl vinyl ether[3]. It has been used as synthetic building block[4][5].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Asymmetry, 18, 557-557 (2007)
B Landfald et al.
Journal of bacteriology, 165(3), 849-855 (1986-03-01)
Glycine betaine and its precursors choline and glycine betaine aldehyde have been found to confer a high level of osmotic tolerance when added exogenously to cultures of Escherichia coli at an inhibitory osmotic strength. In this paper, the following findings
Il Jung et al.
The Journal of organic chemistry, 72(10), 3652-3658 (2007-03-31)
New organic dyes composed of the benzo[b]furan donor, thiophene-conjugated bridge, and cyano acrylic acid acceptor have been newly synthesized through the one-pot coupling cyclization key step. Nanocrystalline TiO2 dye-sensitized solar cell was fabricated using this dye. A solar-to-electric conversion efficiency
Synthesis of graft copolymers based on selective living cationic polymerization using an acetal group with a combination of Lewis acids.
Shimomoto H, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 50(18), 3703-3709 (2012)
Ana Cristina Lima Leite et al.
Bioorganic & medicinal chemistry, 14(11), 3749-3757 (2006-02-07)
A novel series of thiosemicarbazone and aminoacyl-thiazolidones derivatives were synthesized. Their structure suggests that these compounds could have anti-Trypanosoma cruzi activity. Biological evaluation indicates that some of these compounds are able to inhibit the growth of T. cruzi in concentrations

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service