Skip to Content
Merck
All Photos(1)

Key Documents

123943

Sigma-Aldrich

Bis(4-nitrophenyl) phosphate

99%

Synonym(s):

Di-4-nitrophenyl hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2NC6H4O)2P(O)OH
CAS Number:
Molecular Weight:
340.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

solid

mp

172-175 °C (lit.)

functional group

nitro
phosphate

SMILES string

OP(=O)(Oc1ccc(cc1)[N+]([O-])=O)Oc2ccc(cc2)[N+]([O-])=O

InChI

1S/C12H9N2O8P/c15-13(16)9-1-5-11(6-2-9)21-23(19,20)22-12-7-3-10(4-8-12)14(17)18/h1-8H,(H,19,20)

InChI key

MHSVUSZEHNVFKW-UHFFFAOYSA-N

Application

Bis(4-nitrophenyl) phosphate(BNPP) has been used as substrate to determine the enzyme activity of root phosphodiesterases of wetland plants[1]. BNPP has been used to study the mechanism of cleavage of BNPP using oxamido-bridged dinuclear copper(II) complexes as catalysts[2].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M P Lim et al.
Cell death & disease, 2, e170-e170 (2011-06-10)
The major cellular event in the development and progression of liver fibrosis is the activation of hepatic stellate cells (HSCs). Activated HSCs proliferate and produce excess collagen, leading to accumulation of scar matrix and fibrotic liver. As such, the induction
Eliska Rejmánková et al.
The New phytologist, 190(4), 968-976 (2011-06-30)
Phosphorus (P)-limited plants produce higher amounts of root phosphatases, but research has mostly focused on phosphomonoesterases (PMEs). Because phosphate diesters can form a significant proportion of organic P in wetlands, we aimed to determine whether wetland plants produce both root
Studies on the reaction kinetics and the mechanism of hydrolysis of bis (4-nitrophenyl) phosphate (BNPP) catalyzed by oxamido-bridged dinuclear copper (II) complexes in micellar solution.
Xie J, et al.
Transition Met. Chem. (London), 28(7), 782-787 (2003)
Wael M Abdel-Mageed et al.
Plants (Basel, Switzerland), 10(1) (2021-01-13)
A new lupane caffeoyl ester, lup-20(29)-ene 3β-caffeate-30-al (7), and a new oleanane-type triterpene, 3β-hydroxyolean-13(18)-en-12-one (17), were isolated from the aerial parts of Dobera glabra (Forssk), along with ten known triterpenes, including seven lupane-type lupeol (1), 30-nor-lup-3β-ol-20-one (2), ∆1-lupenone (3), lup-20(29)-en-3β,30-diol
Sarah Schmitz et al.
Molecular biotechnology, 62(1), 67-78 (2019-11-22)
Non-specific nuclease (NSN) can be applied in industrial downstream processing to remove nucleic acids from crude protein extracts or in cell-sorting systems to degrade nucleic acids derived from lysed cells. PsNuc from the ice-nucleating bacterium Pseudomonas syringae has the ability

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Could you please assist me with selecting a general esterase inhibitor? My application involves a compound that is not stable in plasma and the instability can be caused by esterase.

    1 answer
    1. A potential product of interest for inhibiting plasma esterase enzyme activity to maintain the stability of substances susceptible to esterase degradation is the product Bis(4-nitrophenyl) phosphate (123943).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service