Skip to Content
Merck
All Photos(1)

Key Documents

123188

Sigma-Aldrich

5-Aminovaleric acid

97%, for peptide synthesis

Synonym(s):

5-AVA, 5-Aminopentanoic acid, Homopiperidinic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$86.90
25 G
$265.00

$86.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$86.90
25 G
$265.00

About This Item

Linear Formula:
NH2(CH2)4CO2H
CAS Number:
Molecular Weight:
117.15
Beilstein:
906833
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$86.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

5-Aminovaleric acid, 97%

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

158-161 °C (lit.)

application(s)

peptide synthesis

SMILES string

NCCCCC(O)=O

InChI

1S/C5H11NO2/c6-4-2-1-3-5(7)8/h1-4,6H2,(H,7,8)

InChI key

JJMDCOVWQOJGCB-UHFFFAOYSA-N

Gene Information

human ... SLC15A1(6564)

Looking for similar products? Visit Product Comparison Guide

Application

5-Aminovaleric acid (5-AVA) is used:
  • In the preparation of (5-AVA)x(MA)1-xPbI3, a perovskite for fabricating printable mesoscopic perovskite solar cell.[1]
  • As a spacer in the synthesis of rhenium and technetium-99m labeled insulin.[2]
  • To synthesize dipeptides that self-assemble to form nanotubes in the solid state as well as in solution over a wide range of pH.[3]
  • As a starting material in the total synthesis of an alkaloid, lycoposerramine Z.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A hole-conductor?free, fully printable mesoscopic perovskite solar cell with high stability.
Mei A, et al.
Science, 345(6194), 295-298 (2014)
cis-Decahydroquinolines via asymmetric organocatalysis: Application to the total synthesis of lycoposerramine Z.
Bradshaw B, et al.
Organic Letters, 15(2), 326-329 (2012)
Ariel Kwaka et al.
Molecular pharmacology, 94(5), 1289-1297 (2018-09-09)
Nematodes exhibit a vast array of cys-loop ligand-gated ion channels with unique pharmacologic characteristics. However, many of the structural components that govern the binding of various ligands are unknown. The nematode cys-loop GABA receptor uncoordinated 49 (UNC-49) is an important
Synthesis and characterization of rhenium and technetium-99m labeled insulin.
Sundararajan C, et al.
Journal of Medicinal Chemistry, 53(6), 2612-2621 (2010)
J C Fothergill et al.
Journal of general microbiology, 99(1), 139-155 (1977-03-01)
Pseudomonas aeruginosa PACI grows poorly on L-lysine as sole source of carbon but mutant derivatives which grow rapidly were readily isolated. Studies with one such mutant, P. aeruginosa PAC586, supported the existence of a route for L-lysine catabolism which differes

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service