Skip to Content
Merck
All Photos(1)

Documents

121983

Sigma-Aldrich

α-(4-Pyridyl)benzhydrol

75%, technical grade

Synonym(s):

α,α-Diphenyl-4-pyridinemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15NO
CAS Number:
Molecular Weight:
261.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

Assay

75%

SMILES string

OC(c1ccccc1)(c2ccccc2)c3ccncc3

InChI

1S/C18H15NO/c20-18(15-7-3-1-4-8-15,16-9-5-2-6-10-16)17-11-13-19-14-12-17/h1-14,20H

InChI key

MRHLFZXYRABVOZ-UHFFFAOYSA-N

General description

α-(4-Pyridyl)benzhydrol(LOH) acts as a ligand and reacts with zinc halides giving the complexes [Zn(LOH)2X2] (X = Cl; X = Br; X = I). It reacts with Ag(CF3SO3) to form a mixed organic/inorganic analogue of wheel-and-axle diol complex [Ag(LOH)2(CF3SO3)(CH3CN)].

Application

Thiol-protecting reagent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Structural Role of the Triflate Anion in the Non-Covalent Silver Polymer [Ag (LOH)2 (CF3SO3)(CH3CN)]{LOH= a-(4-Pyridyl) benzhydrol}.
Bacchi A, et al.
European Journal of Inorganic Chemistry, 2004(10), 1985-1991 (2004)
Unusual hydrogen bonded (OH) 4 tetrahedral nests organize zinc (II) coordination complexes in a non covalent diamondoid network.
Bacchi A, et al.
CrystEngComm, 9(4), 313-318 (2007)
Silke B Bodendiek et al.
Frontiers in pharmacology, 3, 106-106 (2012-06-12)
The paucity of specific pharmacological agents has been a major impediment for delineating the roles of gap junction (GJ) channels formed by connexin proteins in physiology and pathophysiology. Here, we used the selective optimization of side activities (SOSA) approach, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service