Skip to Content
Merck
All Photos(3)

Key Documents

117498

Sigma-Aldrich

1,2-Diaminopropane

1,2-Diaminopropane

99%

Synonym(s):

1,2-Propanediamine, Propylenediamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$37.50
500 G
$103.00
2 KG
$345.00

$37.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$37.50
500 G
$103.00
2 KG
$345.00

About This Item

Linear Formula:
CH3CH(NH2)CH2NH2
CAS Number:
Molecular Weight:
74.12
Beilstein:
605274
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39030209
PubChem Substance ID:
NACRES:
NA.22

$37.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

2.6 (vs air)

Quality Level

vapor pressure

14 mmHg ( 20 °C)

Assay

99%

autoignition temp.

680 °F

expl. lim.

11.1 %

refractive index

n20/D 1.446 (lit.)

bp

119-120 °C (lit.)

solubility

H2O: very soluble

density

0.87 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CC(N)CN

InChI

1S/C3H10N2/c1-3(5)2-4/h3H,2,4-5H2,1H3

InChI key

AOHJOMMDDJHIJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Diaminopropane undergoes condensation with salicylaldehyde and its derivatives to form Schiff base[1]. It reacts with formaldehyde and hydrogen peroxide in aqueous acidic solution to give cage peroxide, 1,6-diaza-3,4,8,9-tetraoxa-11-methylbicyclo[4.4.2]dodecane[2].

Application

1,2-Diaminopropane was used as model precursor in the electron induced deposition of amorphous carbon nitride films[3].

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Flam. Liq. 3 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chi Chen et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-16)
Utilizing liquid rubber to toughen epoxy resin is one of the most mature and promising methods. However, the dielectric relaxation characteristics of the epoxy/liquid rubber composites have not been studied systematically, while the relaxation behaviours are a critical factor for
Ekaterina Yu Bezsudnova et al.
Applied microbiology and biotechnology, 102(22), 9621-9633 (2018-09-05)
Substrate and reaction promiscuity is a remarkable property of some enzymes and facilitates the adaptation to new metabolic demands in the evolutionary process. Substrate promiscuity is also a basis for protein engineering for biocatalysis. However, molecular principles of enzyme promiscuity
Growth and Microstructure of Nanoscale Amorphous Carbon Nitride Films Deposited by Electron Beam Irradiation of 1, 2-Diaminopropane.
Wnuk JD, et al.
The Journal of Physical Chemistry C, 113(28), 12345-12354 (2009)
Mononuclear and dinuclear chiral vanadium (V) complexes with tridentate Schiff bases derived from< i> R</i>(-)-1, 2-diaminopropane: Synthesis, structure, characterization and catalytic properties.
Romanowski G and Wera M.
Polyhedron, 29(13), 2747-2754 (2010)
Maryam Ghiyasiyan-Arani et al.
Scientific reports, 6, 25231-25231 (2016-05-05)
In this work, copper pyrovanadate (Cu3V2O7(OH)2(H2O)2) nanoparticles have been synthesized by a simple and rapid chemical precipitation method. Different copper-organic complexes were used to control the size and morphology of products. The morphology and structure of the as-synthesized products were

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service