Skip to Content
Merck
All Photos(1)

Key Documents

114774

Sigma-Aldrich

3′,3′′,5′,5′′-Tetrabromophenolphthalein ethyl ester potassium salt

indicator grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H13Br4KO4
CAS Number:
Molecular Weight:
700.05
Beilstein:
3848263
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

indicator grade

visual transition interval

3.0-3.5, yellow to green
4.2-4.9, blue-green to blue-violet

mp

210 °C (dec.) (lit.)

λmax

386 nm (2nd)
593 nm

SMILES string

[K+].CCOC(=O)c1ccccc1\C(c2cc(Br)c([O-])c(Br)c2)=C3\C=C(Br)C(=O)C(Br)=C3

InChI

1S/C22H14Br4O4.K/c1-2-30-22(29)14-6-4-3-5-13(14)19(11-7-15(23)20(27)16(24)8-11)12-9-17(25)21(28)18(26)10-12;/h3-10,27H,2H2,1H3;/q;+1/p-1

InChI key

WCIQBKUTYDIBJC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

3′,3′′,5′,5′′-Tetrabromophenolphthalein ethyl ester is used in the spectrophotometric method of total protein determination.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric determination of total proteins in blood plasma: a comparative study among dye-binding methods.
Zaia DAM, et al.
Brazilian Archives of Biology and Technology, 48, 385-388 (2005)
[Pharmaco-analytical application of tetrabromophenolphthalein ethyl ester indicator dye. III. Interaction between alkaloids and TBPE].
G Szász et al.
Acta pharmaceutica Hungarica, 50(4), 161-165 (1980-07-01)
H T Chang et al.
Electrophoresis, 22(3), 394-398 (2001-03-22)
Electroelution of protein bands from a gel has advantages over the competitive common technique requiring gel sectioning with respect to yield, speed and the potential for computer-controlled application to multicomponent two-dimensional (2-D) gels. The electroelution design for the commercial high-performance
The application of the TBP test to bile.
E G Saker
Journal of analytical toxicology, 8(1), 51-51 (1984-01-01)
[Application of tetrabromophenolphthalein ethyl ester (TBPE) indicator-dye to pharmaco-analysis].
G Szász et al.
Acta pharmaceutica Hungarica, 51(1), 43-47 (1981-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service