Skip to Content
Merck
All Photos(2)

Key Documents

113212

Sigma-Aldrich

1-Phenyldecane

98%

Synonym(s):

Decylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$305.00
100 G
$1,190.00

$305.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$305.00
100 G
$1,190.00

About This Item

Linear Formula:
C6H5(CH2)9CH3
CAS Number:
Molecular Weight:
218.38
Beilstein:
1907745
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$305.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

refractive index

n20/D 1.482 (lit.)

bp

293 °C (lit.)

mp

−14 °C (lit.)

density

0.856 g/mL at 25 °C (lit.)

functional group

phenyl

SMILES string

CCCCCCCCCCc1ccccc1

InChI

1S/C16H26/c1-2-3-4-5-6-7-8-10-13-16-14-11-9-12-15-16/h9,11-12,14-15H,2-8,10,13H2,1H3

InChI key

UZILCZKGXMQEQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Decylbenzene (1-Phenyldecane) forms a charge-transfer complex with fluoranil.

Application

1-Phenyldecane is used as an enhancer to reduce interference in ion-selective membrane electrodes for determining carbon dioxide[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W J Scott et al.
Clinical chemistry, 32(1 Pt 1), 137-141 (1986-01-01)
Previously described ion-selective membrane electrodes useful for determination of total carbon dioxide in biological fluids in automated analyzers show interference from several sources--e.g., fatty acids, keto acids, salicylate, heparin. Judicious selection of membrane components has produced a membrane with superior
K Elias Ikonen et al.
Journal of chromatography. A, 1614, 460705-460705 (2019-11-23)
An update for ASTM D2425, "Standard Test Method for Hydrocarbon Types in Middle Distillates by Mass Spectrometry," was recently released to expand its scope by adding modern mass spectrometric instrumentation and a new sample type, synthesized hydrocarbons. While the method

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service