Skip to Content
Merck
All Photos(1)

Key Documents

112380

Sigma-Aldrich

Benzyl salicylate

98%

Synonym(s):

Benzyl 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2CH2C6H5
CAS Number:
Molecular Weight:
228.24
Beilstein:
2115365
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39024514
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.581 (lit.)

bp

168-170 °C/5 mmHg (lit.)

density

1.176 g/mL at 25 °C (lit.)

SMILES string

Oc1ccccc1C(=O)OCc2ccccc2

InChI

1S/C14H12O3/c15-13-9-5-4-8-12(13)14(16)17-10-11-6-2-1-3-7-11/h1-9,15H,10H2

InChI key

ZCTQGTTXIYCGGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl salicylate is a constituent of raw propolis that can be identified by headspace solid-phase microextraction (HS-SPME), followed by gas chromatography-mass spectrometry (GC-MS)[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

349.7 °F - closed cup

Flash Point(C)

176.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A H Gold et al.
Molecular and cellular biochemistry, 25(1), 47-59 (1979-05-06)
Liver glycogen synthase b phosphatase, chromatographically separable from phosphorylase a phosphatase, is decreased in 48-hour alloxan diabetic rats. The phosphatase activities are measured in an in vitro system using exogenous isolated phospho-enzyme as substrates with added phosphatases. Synthase and phosphorylase
Marcel Holzer et al.
Archiv der Pharmazie, 341(8), 478-484 (2008-07-12)
Starting point of the present paper was the result of a virtual screening using the open conformation of the large extracellular loop (LEL) of the CD81-receptor (crystal structure: PDB-ID: 1G8Q). After benzyl salicylate had been experimentally validated to be a
A Lapczynski et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45 Suppl 1, S362-S380 (2007-11-24)
A toxicologic and dermatologic review of benzyl salicylate when used as a fragrance ingredient is presented.
Erik Christian Munksgaard
European journal of oral sciences, 112(1), 101-104 (2004-02-12)
The loss of the plasticizers dibutylphthalate, butylphthalylbutyl glycolate, benzylbenzoate, methylsalicylate, and benzylsalicylate from four soft lining materials was measured. A 0.1% aqueous solution of triton X-100, reduced was used as immersion medium, since the solubility of plasticizer in this medium
D W Jones et al.
Journal of dental research, 65(5), 634-642 (1986-05-01)
An evaluation has been made of the soft polymer gel systems of the type used as prosthodontic short-term denture-lining materials. The purpose of the study was to determine the effect upon gel strength and gelation time produced by variations in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service