Skip to Content
Merck
All Photos(1)

Key Documents

10911

Sigma-Aldrich

tert-Butyl acetoacetate

produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0% (GC)

Synonym(s):

ACTB

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€563.70
100 MG
€723.60
250 MG
€1,017.40
500 MG
€1,576.40
1 G
€2,008.00
2.5 G
€3,894.30
5 G
€5,739.00
10 G
€8,487.20

Vendor SKU: EN300-247645-50MG

€563.70


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
€563.70
100 MG
€723.60
250 MG
€1,017.40
500 MG
€1,576.40
1 G
€2,008.00
2.5 G
€3,894.30
5 G
€5,739.00
10 G
€8,487.20

About This Item

Linear Formula:
CH3COCH2COOC(CH3)3
CAS Number:
Molecular Weight:
158.19
Beilstein:
1680303
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Vendor SKU: EN300-247645-50MG

€563.70


Ships in 14 days - from Aldrich Partner.

grade

produced by Wacker Chemie AG, Burghausen, Germany

Assay

≥98.0% (GC)

manufacturer/tradename

Wacker Chemie AG

refractive index

n20/D 1.419 (lit.)

bp

71-72 °C/11 mmHg (lit.)

density

0.954 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CC(=O)OC(C)(C)C

InChI

1S/C8H14O3/c1-6(9)5-7(10)11-8(2,3)4/h5H2,1-4H3

InChI key

JKUYRAMKJLMYLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

prices for bulk quantities on request

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meghan C Drummond et al.
Nature communications, 6, 6873-6873 (2015-04-22)
The maintenance of sensory hair cell stereocilia is critical for lifelong hearing; however, mechanisms of structural homeostasis remain poorly understood. Conflicting models propose that stereocilia F-actin cores are either continually renewed every 24-48 h via a treadmill or are stable, exceptionally
Hong Xie et al.
Oncotarget, 5(20), 10048-10057 (2014-10-11)
Telomerase activation through the induction of its catalytic component TERT is essential in carcinogenesis. The regulatory mechanism and clinical significance underlying cancer-specific TERT expression have been extensively investigated in various human malignancies, but little is known about these in Merkel
Jordan M Spatz et al.
The Journal of biological chemistry, 290(27), 16744-16758 (2015-05-09)
Although bone responds to its mechanical environment, the cellular and molecular mechanisms underlying the response of the skeleton to mechanical unloading are not completely understood. Osteocytes are the most abundant but least understood cells in bones and are thought to
Ying Wang et al.
Endocrinology, 155(5), 1970-1981 (2014-03-08)
FSH is an essential regulator of mammalian reproduction. Its synthesis by pituitary gonadotrope cells is regulated by multiple endocrine and paracrine factors, including TGFβ superfamily ligands, such as the activins and inhibins. Activins stimulate FSH synthesis via transcriptional regulation of
Hiroshi Ono et al.
Cancer medicine, 3(3), 632-642 (2014-03-05)
Small-cell lung cancer (SCLC) is a subtype of lung cancer with poor prognosis. To identify accurate predictive biomarkers and effective therapeutic modalities, we focus on a long noncoding RNA, Hox transcript antisense intergenic RNA (HOTAIR), and investigated its expression, cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service