Skip to Content
Merck
All Photos(2)

Documents

108073

Sigma-Aldrich

8-Hydroxyquinoline hemisulfate salt hemihydrate

98%

Synonym(s):

8-Quinolinol hemisulfate, 8-Quinolinol sulfate (2:1) monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO · 0.5H2O4S · 0.5H2O
CAS Number:
Molecular Weight:
203.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

176-179 °C (lit.)

solubility

H2O: soluble 100 mg/mL, clear to slightly hazy, yellow to orange

SMILES string

[H]O[H].OS(O)(=O)=O.Oc1cccc2cccnc12.Oc3cccc4cccnc34

InChI

1S/2C9H7NO.H2O4S.H2O/c2*11-8-5-1-3-7-4-2-6-10-9(7)8;1-5(2,3)4;/h2*1-6,11H;(H2,1,2,3,4);1H2

InChI key

BNCXJZDIJIVJJO-UHFFFAOYSA-N

Biochem/physiol Actions

It is a potential copper ligand and is a tested effective proteasome inhibitor.

Preparation Note

8-Hydroxyquinoline hemisulfate salt hemihydrate dissolves in water at a concentration of 100 mg/ml to form a clear to slightly hazy, yellow to yellow-orange coloured solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenyon G Daniel et al.
Biochemical pharmacology, 67(6), 1139-1151 (2004-03-10)
Here we report that organic copper complexes can potently and selectively inhibit the chymotrypsin-like activity of the proteasome in vitro and in vivo. Several copper compounds, such as NCI-109268 and bis-8-hydroxyquinoline copper(II) [Cu(8-OHQ)(2)], can inhibit the chymotrypsin-like activity of purified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service